Gene Transcripts

Gene Symbol
HIKESHI
Entrez Gene ID
51501
Gene Name
Hikeshi, heat shock protein nuclear import factor
Gene Aliases
C11orf73, HLD13, HSPC138, HSPC179, L7RN6, OPI10
Chromosome Location
Chr. 11: 86302211 - 86345943 on Build GRCh38
UniGene ID
Hs.283322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322404.1 NP_001309333.1 5 848
NM_001322407.1 NP_001309336.1 6 1082
NM_001322409.1 NP_001309338.1 6 1012
NM_016401.3 NP_057485.2 5 924
NR_024597.1 5 791
NR_024598.1 4 686
NR_136324.1 6 1032
XM_017017915.1 XP_016873404.1 5 701
GenBank mRNA BC015991.2 - 5 889
BC021621.1 - 5 889 2820691

View Full Product Details