Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164835.1 | NP_001158307.1 | 4 | 1197 | |
NM_019079.4 | NP_061952.3 | 3 | 1086 | ||
GenBank mRNA | AB211065.1 | - | 3 | 1024 | |
AK001746.1 | - | 1 | 455 | ||
AK001973.1 | - | 3 | 1012 | ||
AL834314.1 | - | 3 | 1024 | ||
BC111559.1 | - | 3 | 999 | 40035707 |