Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020348.2 | NP_065081.2 | 5 | 2352 | |
XM_005269692.3 | XP_005269749.2 | 5 | 2352 | ||
XM_011539629.1 | XP_011537931.1 | 5 | 2352 | ||
XM_011539630.1 | XP_011537932.1 | 5 | 2352 | ||
XM_011539631.2 | XP_011537933.1 | 5 | 2352 | ||
GenBank mRNA | AF169226.1 | - | 5 | 1175 | |
AK056194.1 | - | 4 | 455 | ||
AK296545.1 | - | 5 | 1520 | ||
AK299346.1 | - | 5 | 1509 | ||
AK308326.1 | - | 4 | 399 | ||
AK308618.1 | - | 5 | 2093 | ||
AK310486.1 | - | 4 | 607 | ||
BC098103.1 | - | 5 | 1126 | 40006972 | |
BC098134.1 | - | 5 | 1126 | ||
BC098279.1 | - | 5 | 1126 | 40006977 | |
BC098307.1 | - | 5 | 1126 | 40006978 | |
JF267782.1 | - | 3 | 282 |