Gene Transcripts

Gene Symbol
MSANTD2
Entrez Gene ID
79684
Gene Name
Myb/SANT DNA binding domain containing 2
Gene Aliases
C11orf61
Chromosome Location
Chr. 11: 124766492 - 124800879 on Build GRCh38
UniGene ID
Hs.146079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308027.1 NP_001294956.1 2 718
NM_001312920.1 NP_001299849.1 1 1452
NM_001312921.1 NP_001299850.1 2 575
NM_024631.3 NP_078907.2 2 562
XM_005271674.3 XP_005271731.3 3 1127
XM_006718905.1 XP_006718968.1 2 2087
XM_006718906.3 XP_006718969.1 2 1923
XM_011542981.2 XP_011541283.2 2 1193
XM_011542982.2 XP_011541284.2 2 1193
XM_017018295.1 XP_016873784.1 2 17333
XM_017018296.1 XP_016873785.1 2 2243
XM_017018297.1 XP_016873786.1 3 544
XM_017018298.1 XP_016873787.1 3 13304
XM_017018299.1 XP_016873788.1 3 345
GenBank mRNA AK026995.1 - 2 561
AK092413.1 - 1 1126
AL512691.1 - 1 1452
BC064919.1 - 2 703 6093009
CX781748.1 - 2 561
DA379833.1 - 2 562

View Full Product Details