Gene Transcripts
- Gene Symbol
- FLAD1
- Entrez Gene ID
- 80308
- Gene Name
- flavin adenine dinucleotide synthetase 1
- Gene Aliases
- FAD1, FADS, LSMFLAD, PP591
- Chromosome Location
- Chr. 1: 154983294 - 154993111 on Build GRCh38
- UniGene ID
- Hs.118666
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025207.4 |
NP_079483.3
|
6, 7 |
2032 |
|
NM_201398.2 |
NP_958800.1
|
7, 8 |
1588 |
|
GenBank mRNA |
AF481877.1 |
- |
6, 7 |
1340 |
|
AF520568.1 |
- |
6, 7 |
1953 |
|
AK025979.1 |
- |
7, 8 |
1516 |
|
BC020253.1 |
- |
5, 6 |
867 |
|
BC021096.1 |
- |
7, 8 |
1506 |
4891331 |
BC032323.1 |
- |
7, 8 |
1523 |
5242727 |
DQ458779.1 |
- |
6, 7 |
1631 |
|
U79241.1 |
- |
5, 6 |
1183 |
|
View Full Product Details