Gene Transcripts

Gene Symbol
HMCN1
Entrez Gene ID
83872
Gene Name
hemicentin 1
Gene Aliases
ARMD1, FBLN6, FIBL-6, FIBL6
Chromosome Location
Chr. 1: 185734526 - 186190953 on Build GRCh38
UniGene ID
Hs.58877
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031935.2 NP_114141.2 7 1154
XM_011510038.2 XP_011508340.1 7 1171
XM_011510041.2 XP_011508343.1 7 1179
GenBank mRNA AF156100.1 - 7 1154

View Full Product Details