Gene Transcripts

Gene Symbol
STK40
Entrez Gene ID
83931
Gene Name
serine/threonine kinase 40
Gene Aliases
SHIK, SgK495
Chromosome Location
Chr. 1: 36339619 - 36385927 on Build GRCh38
UniGene ID
Hs.471768
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282546.1 NP_001269475.1 8 1097
NM_001282547.1 NP_001269476.1 8 1082
NM_032017.2 NP_114406.1 9 1319
GenBank mRNA AK024504.1 - 7 881
AK075029.1 - 5 598
AK075048.1 - 8 1076
AK131560.1 - 1 1459
AL834137.1 - 9 1276
AY198395.1 - 8 1052
BC005169.2 - 3 281
BC007835.2 - 8 1050 4109652
BC008344.1 - 8 1058 3530483

View Full Product Details