Gene Transcripts

Gene Symbol
SRSF8
Entrez Gene ID
10929
Gene Name
serine and arginine rich splicing factor 8
Gene Aliases
DSM-1, SFRS2B, SRP46
Chromosome Location
Chr. 11: 95066877 - 95071227 on Build GRCh38
UniGene ID
Hs.476680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032102.3 NP_115285.1 1 1682
GenBank mRNA AF031166.1 - 1 1302
AK309776.1 - 3 994
BC057783.1 - 2 1372 5285882

View Full Product Details