Gene Transcripts

Gene Symbol
MAB21L3
Entrez Gene ID
126868
Gene Name
mab-21 like 3
Gene Aliases
C1orf161
Chromosome Location
Chr. 1: 116072744 - 116135240 on Build GRCh38
UniGene ID
Hs.376194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152367.2 NP_689580.2 7 2940
GenBank mRNA AK096035.1 - 7 2940

View Full Product Details