Gene Transcripts

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
Chromosome Location
Chr. 1: 235440654 - 235504481 on Build GRCh38
UniGene ID
Hs.498143
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152490.4 NP_689703.1 12 1783
XM_017000394.1 XP_016855883.1 13 1906
XM_017000395.1 XP_016855884.1 13 1870
GenBank mRNA AB209046.1 - 11 1337
BC029564.1 - 12 1783 4826983

View Full Product Details