Gene Transcripts

Gene Symbol
C11orf65
Entrez Gene ID
160140
Gene Name
chromosome 11 open reading frame 65
Gene Aliases
Chromosome Location
Chr. 11: 108307344 - 108469832 on Build GRCh38
UniGene ID
Hs.653180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152587.3 NP_689800.3 3 220
XM_005271412.3 XP_005271469.1 3 236
XM_005271414.3 XP_005271471.1 3 236
XM_005271415.4 XP_005271472.1 3 236
XM_011542639.2 XP_011540941.1 4 273
XM_011542640.2 XP_011540942.1 3 236
XM_011542642.1 XP_011540944.1 3 236
XM_011542643.2 XP_011540945.1 3 236
XM_017017246.1 XP_016872735.1 3 180
XM_017017247.1 XP_016872736.1 3 236
XM_017017248.1 XP_016872737.1 3 236
GenBank mRNA BC029536.1 - 3 193

View Full Product Details