Gene Transcripts

Gene Symbol
MTMR11
Entrez Gene ID
10903
Gene Name
myotubularin related protein 11
Gene Aliases
CRA
Chromosome Location
Chr. 1: 149928651 - 149936898 on Build GRCh38
UniGene ID
Hs.425144
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145862.1 NP_001139334.1 5 620
NM_181873.3 NP_870988.2 4 362
XM_006711137.1 XP_006711200.1 4 339
XM_011509099.2 XP_011507401.1 5 513
GenBank mRNA AK097000.1 - 4 339
AK296094.1 - 4 459
AK298775.1 - 5 589
AK304691.1 - 4 436
AK304820.1 - 5 590
BC032655.1 - 4 301
BC064372.1 - 5 592
BC094756.1 - 5 601
BC100818.2 - 4 280
BC100819.2 - 4 280
BC100820.1 - 4 280
BC100821.2 - 4 280
U78556.1 - 5 555
U78557.1 - 5 555

View Full Product Details