Gene Transcripts

Gene Symbol
FAAP20
Entrez Gene ID
199990
Gene Name
Fanconi anemia core complex associated protein 20
Gene Aliases
C1orf86, FP7162
Chromosome Location
Chr. 1: 2184460 - 2212720 on Build GRCh38
UniGene ID
Hs.107101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282671.1 NP_001269600.1 6 748
NM_001282672.1 NP_001269601.1 5 617
XM_017000555.1 XP_016856044.1 1 196
XM_017000556.1 XP_016856045.1 1 196
GenBank mRNA AF469129.1 - 6 714
AK055593.1 - 5 617
BC103992.2 - 5 617 40022365

View Full Product Details