Gene Transcripts

Gene Symbol
MDFIC
Entrez Gene ID
29969
Gene Name
MyoD family inhibitor domain containing
Gene Aliases
HIC
Chromosome Location
Chr. 7: 114922154 - 115019916 on Build GRCh38
UniGene ID
Hs.741413
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166345.1 NP_001159817.1 5 4056
NM_199072.4 NP_951038.1 5 4056
GenBank mRNA AF054589.1 - 5 4060

View Full Product Details