Gene Transcripts
- Gene Symbol
- IGF2BP2
- Entrez Gene ID
- 10644
- Gene Name
- insulin like growth factor 2 mRNA binding protein 2
- Gene Aliases
- IMP-2, IMP2, VICKZ2
- Chromosome Location
- Chr. 3: 185643739 - 185825051 on Build GRCh38
- UniGene ID
- Hs.35354
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001007225.1 |
NP_001007226.1
|
3, 4 |
375 |
|
NM_001291869.1 |
NP_001278798.1
|
3, 4 |
375 |
|
NM_001291872.1 |
NP_001278801.1
|
2, 3 |
177 |
|
NM_001291873.1 |
NP_001278802.1
|
2, 3 |
177 |
|
NM_001291874.1 |
NP_001278803.1
|
2, 3 |
177 |
|
NM_001291875.1 |
NP_001278804.1
|
2, 3 |
177 |
|
NM_006548.4 |
NP_006539.3
|
3, 4 |
375 |
|
XM_017005557.1 |
XP_016861046.1
|
2, 3 |
320 |
|
XM_017005558.1 |
XP_016861047.1
|
3, 4 |
380 |
|
XM_017005559.1 |
XP_016861048.1
|
3, 4 |
381 |
|
XM_017005560.1 |
XP_016861049.1
|
3, 4 |
277 |
|
XM_017005561.1 |
XP_016861050.1
|
3, 4 |
382 |
|
XM_017005562.1 |
XP_016861051.1
|
3, 4 |
381 |
|
XM_017005563.1 |
XP_016861052.1
|
3, 4 |
381 |
|
XM_017005564.1 |
XP_016861053.1
|
3, 4 |
381 |
|
GenBank mRNA |
AF117107.1 |
- |
3, 4 |
342 |
|
AK290664.1 |
- |
3, 4 |
376 |
|
AK309611.1 |
- |
3, 4 |
329 |
|
BC021290.2 |
- |
3, 4 |
360 |
5090334 |
EU408701.1 |
- |
2, 3 |
177 |
|
EU408702.1 |
- |
2, 3 |
177 |
|
EU408703.1 |
- |
2, 3 |
177 |
|
EU408704.1 |
- |
2, 3 |
177 |
|
View Full Product Details