Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014306.4 | NP_055121.1 | 4 | 410 | |
GenBank mRNA | AF155658.1 | - | 4 | 358 | |
AF161466.1 | - | 4 | 369 | ||
AK297908.1 | - | 370 | |||
AK301945.1 | - | 4 | 410 | ||
AK312503.1 | - | 4 | 370 | ||
AL050255.1 | - | 4 | 354 | ||
BC000151.1 | - | 4 | 348 | 2984824 | |
BC002970.1 | - | 4 | 348 | 3533883 | |
BC010308.1 | - | 4 | 360 | 3451576 | |
BC016707.1 | - | 4 | 375 | 4102148 | |
CR456450.1 | - | 4 | 325 | ||
CR457175.1 | - | 4 | 279 | ||
DC396330.1 | - | 4 | 412 |