Gene Transcripts

Gene Symbol
PID1
Entrez Gene ID
55022
Gene Name
phosphotyrosine interaction domain containing 1
Gene Aliases
HMFN2073, NYGGF4, P-CLI1, PCLI1
Chromosome Location
Chr. 2: 229023973 - 229271341 on Build GRCh38
UniGene ID
Hs.745038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017933.4 NP_060403.3 2, 3 536
XM_017004404.1 XP_016859893.1 2, 3 5467
GenBank mRNA AK125359.1 - 2, 3 460

View Full Product Details