Gene Transcripts

Gene Symbol
PTCD3
Entrez Gene ID
55037
Gene Name
pentatricopeptide repeat domain 3
Gene Aliases
MRP-S39
Chromosome Location
Chr. 2: 86106182 - 86142157 on Build GRCh38
UniGene ID
Hs.323489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017952.5 NP_060422.4 6, 7 477
GenBank mRNA BC011832.2 - 6, 7 412
BI825315.1 - 6, 7 464

View Full Product Details