Gene Transcripts

Gene Symbol
BAIAP2L2
Entrez Gene ID
80115
Gene Name
BAI1 associated protein 2 like 2
Gene Aliases
Chromosome Location
Chr. 22: 38084889 - 38110944 on Build GRCh38
UniGene ID
Hs.474822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025045.5 NP_079321.3 6 512
XM_005261751.4 XP_005261808.1 7 513
XM_011530379.2 XP_011528681.1 6 629
XM_011530380.2 XP_011528682.1 5 415
XM_011530381.2 XP_011528683.1 6 406
XM_011530382.2 XP_011528684.1 6 794
XM_011530383.2 XP_011528685.1 5 732
XM_011530384.2 XP_011528686.1 6 670
GenBank mRNA AK026235.1 - 6 470
AY358164.1 - 6 512

View Full Product Details