Gene Transcripts
- Gene Symbol
- SMARCD2
- Entrez Gene ID
- 6603
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 2
- Gene Aliases
- BAF60B, CRACD2, PRO2451, Rsc6p
- Chromosome Location
- Chr. 17: 63832081 - 63842991 on Build GRCh38
- UniGene ID
- Hs.250581
- Species
- Homo sapiens
- Gene Symbol
- PTCRA
- Entrez Gene ID
- 171558
- Gene Name
- pre T-cell antigen receptor alpha
- Gene Aliases
- PT-ALPHA, PTA
- Chromosome Location
- Chr. 6: 42915989 - 42925838 on Build GRCh38
- UniGene ID
- Hs.169002
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130819.1 |
- |
|
995 |
|
View Full Product Details