Gene Transcripts

Gene Symbol
NDUFAF7
Entrez Gene ID
55471
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Gene Aliases
C2orf56, MidA, PRO1853
Chromosome Location
Chr. 2: 37231631 - 37259052 on Build GRCh38
UniGene ID
Hs.433466
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083946.1 NP_001077415.1 2 275
NM_144736.4 NP_653337.1 2 275
XM_005264410.1 XP_005264467.1 2 255
XM_005264411.1 XP_005264468.1 2 253
XM_011532956.1 XP_011531258.1 2 243
GenBank mRNA AK294240.1 - 2 209
AK299015.1 - 2 249
AK302150.1 - 2 226
AK303555.1 - 2 255
BC004548.2 - 2 240 3959139
BI334629.1 - 2 226
BM172474.1 - 2 280
BX538031.1 - 2 228

View Full Product Details