Gene Transcripts
- Gene Symbol
- NDUFAF7
- Entrez Gene ID
- 55471
- Gene Name
- NADH:ubiquinone oxidoreductase complex assembly factor 7
- Gene Aliases
- C2orf56, MidA, PRO1853
- Chromosome Location
- Chr. 2: 37231631 - 37259052 on Build GRCh38
- UniGene ID
- Hs.433466
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144736.4 |
NP_653337.1
|
2, 3 |
299 |
|
XM_005264410.1 |
XP_005264467.1
|
2, 3 |
279 |
|
XM_005264411.1 |
XP_005264468.1
|
2, 3 |
277 |
|
XM_011532956.1 |
XP_011531258.1
|
2, 3 |
267 |
|
GenBank mRNA |
AK294240.1 |
- |
2, 3 |
233 |
|
AK299015.1 |
- |
2, 3 |
273 |
|
AK303555.1 |
- |
2, 3 |
279 |
|
BC004548.2 |
- |
2, 3 |
264 |
3959139 |
BI334629.1 |
- |
2, 3 |
250 |
|
BM172474.1 |
- |
2, 3 |
304 |
|
View Full Product Details