Gene Transcripts

Gene Symbol
IQCF1
Entrez Gene ID
132141
Gene Name
IQ motif containing F1
Gene Aliases
Chromosome Location
Chr. 3: 51894876 - 51903370 on Build GRCh38
UniGene ID
Hs.671210
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152397.2 NP_689610.2 2, 3 208
GenBank mRNA AK131018.1 - 2, 3 159
BC029595.1 - 2, 3 173 5273825
BC034228.1 - 2, 3 209

View Full Product Details