Gene Transcripts

Gene Symbol
WDSUB1
Entrez Gene ID
151525
Gene Name
WD repeat, sterile alpha motif and U-box domain containing 1
Gene Aliases
UBOX6, WDSAM1
Chromosome Location
Chr. 2: 159235793 - 159286799 on Build GRCh38
UniGene ID
Hs.20848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128212.2 NP_001121684.1 9 1221
NM_001128213.1 NP_001121685.1 9 1237
NM_001307994.1 NP_001294923.1 5 945
NM_152528.2 NP_689741.2 9 1129
XM_005246355.2 XP_005246412.2 9 1215
XM_006712341.3 XP_006712404.1 8 709
XM_011510753.2 XP_011509055.1 9 1181
XM_011510754.2 XP_011509056.1 9 915
XM_017003481.1 XP_016858970.1 9 1215
XM_017003482.1 XP_016858971.1 4 846
XM_017003483.1 XP_016858972.1 8 1036
GenBank mRNA AK093494.1 - 9 1129
AK129983.1 - 9 1237
AL040518.1 - 8 605
BC029520.1 - 5 849 5301559

View Full Product Details