Gene Transcripts

Gene Symbol
RGL4
Entrez Gene ID
266747
Gene Name
ral guanine nucleotide dissociation stimulator like 4
Gene Aliases
Rgr
Chromosome Location
Chr. 22: 23690861 - 23699176 on Build GRCh38
UniGene ID
Hs.658997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153615.1 NP_705843.1 5 2153
GenBank mRNA AK131340.1 - 7 2591
AY101396.1 - 5 2153
BC101108.3 - 4 1833 40011557
BC101109.2 - 4 1833 40011564
BC101110.3 - 5 1708

View Full Product Details