Gene Transcripts

Gene Symbol
SELV
Entrez Gene ID
348303
Gene Name
selenoprotein V
Gene Aliases
Chromosome Location
Chr. 19: 39515113 - 39520686 on Build GRCh38
UniGene ID
Hs.319284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182704.1 NP_874363.1 6 1628
GenBank mRNA AY324825.1 - 6 1628

View Full Product Details