Gene Transcripts

Gene Symbol
LONRF2
Entrez Gene ID
164832
Gene Name
LON peptidase N-terminal domain and ring finger 2
Gene Aliases
RNF192
Chromosome Location
Chr. 2: 100273291 - 100322733 on Build GRCh38
UniGene ID
Hs.21380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198461.3 NP_940863.3 12 2858
GenBank mRNA AK127206.1 - 12 1560
BC101662.1 - 11 1517 8069168
BC101664.1 - 11 1517 8069170
BC143468.1 - 11 1517
DQ020495.1 - 12 2858

View Full Product Details