Gene Transcripts

Gene Symbol
RABEPK
Entrez Gene ID
10244
Gene Name
Rab9 effector protein with kelch motifs
Gene Aliases
RAB9P40, bA65N13.1, p40
Chromosome Location
Chr. 9: 125200493 - 125234159 on Build GRCh38
UniGene ID
Hs.19012
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174152.1 NP_001167623.1 9 1011
NM_001174153.1 NP_001167624.1 8 858
NM_005833.3 NP_005824.2 8 1172
XM_005251640.4 XP_005251697.1 8 1084
XM_005251641.4 XP_005251698.1 7 939
XM_005251642.4 XP_005251699.1 7 930
XM_005251644.4 XP_005251701.1 6 780
XM_011518120.2 XP_011516422.1 8 1246
XM_017014177.1 XP_016869666.1 9 996
XM_017014178.1 XP_016869667.1 8 898
XM_017014179.1 XP_016869668.1 7 971
XM_017014180.1 XP_016869669.1 7 745
XM_017014181.1 XP_016869670.1 6 856
XM_017014182.1 XP_016869671.1 7 681
XM_017014183.1 XP_016869672.1 7 1177
GenBank mRNA AK290768.1 - 8 1016
BC000503.2 - 8 1149 2821471
BC047023.1 - 8 838 5396980
BC053541.1 - 8 981 5724458
BC065725.1 - 9 1014 4102355
CR456837.1 - 7 842
CR541851.1 - 7 842
KU178507.1 - 7 842
KU178508.1 - 7 755
KU178509.1 - 6 693
KU178510.1 - 6 689
KU178511.1 - 5 527
Z97074.1 - 9 991

View Full Product Details