Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317381.1 | NP_001304310.1 | 4 | 547 | |
NM_001317382.1 | NP_001304311.1 | 3 | 266 | ||
NM_001317383.1 | NP_001304312.1 | 3 | 557 | ||
NM_014278.3 | NP_055093.2 | 3 | 557 | ||
XM_011531745.2 | XP_011530047.1 | 3 | 626 | ||
GenBank mRNA | AB023421.1 | - | 3 | 269 | |
AK302115.1 | - | 3 | 240 | ||
AK303053.1 | - | 4 | 547 | ||
AK307464.1 | - | 3 | 444 | ||
AY290862.1 | - | 3 | 423 | ||
BC040560.1 | - | 3 | 468 | 5270824 | |
DB086945.1 | - | 3 | 266 | ||
EF197155.1 | - | 3 | 215 |