Gene Transcripts
- Gene Symbol
- SRRT
- Entrez Gene ID
- 51593
- Gene Name
- serrate, RNA effector molecule
- Gene Aliases
- ARS2, ASR2, serrate
- Chromosome Location
- Chr. 7: 100874971 - 100888664 on Build GRCh38
- UniGene ID
- Hs.111801
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128852.1 |
NP_001122324.1
|
5, 6 |
858 |
|
NM_001128853.1 |
NP_001122325.1
|
5, 6 |
858 |
|
NM_001128854.1 |
NP_001122326.1
|
5, 6 |
858 |
|
NM_015908.5 |
NP_056992.4
|
5, 6 |
858 |
|
XM_005250405.2 |
XP_005250462.1
|
5, 6 |
715 |
|
XM_005250406.2 |
XP_005250463.1
|
5, 6 |
715 |
|
XM_005250407.2 |
XP_005250464.1
|
5, 6 |
715 |
|
XM_005250408.2 |
XP_005250465.1
|
5, 6 |
715 |
|
XM_017012290.1 |
XP_016867779.1
|
6, 7 |
1040 |
|
XM_017012291.1 |
XP_016867780.1
|
6, 7 |
1040 |
|
GenBank mRNA |
AF248955.1 |
- |
5, 6 |
821 |
|
AL096723.1 |
- |
5, 6 |
813 |
|
BC069249.1 |
- |
3, 4 |
399 |
|
BC109117.1 |
- |
5, 6 |
752 |
40035609 |
BG477711.1 |
- |
|
241 |
|
View Full Product Details