Gene Transcripts

Gene Symbol
PBDC1
Entrez Gene ID
51260
Gene Name
polysaccharide biosynthesis domain containing 1
Gene Aliases
CXorf26
Chromosome Location
Chr. X: 76172929 - 76178310 on Build GRCh38
UniGene ID
Hs.370100
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300888.1 NP_001287817.1 4 460
NM_016500.3 NP_057584.2 4 453
GenBank mRNA BC001220.1 - 4 453
BC051894.1 - 4 315 6528202
BM817702.1 - 4 349
CD704296.1 - 4 354

View Full Product Details