Gene Transcripts

Gene Symbol
TRMT12
Entrez Gene ID
55039
Gene Name
tRNA methyltransferase 12 homolog
Gene Aliases
TRM12, TYW2
Chromosome Location
Chr. 8: 124450807 - 124453026 on Build GRCh38
UniGene ID
Hs.9925
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017956.3 NP_060426.2 1 222
GenBank mRNA AK000779.1 - 1 210
AK222727.1 - 1 210
AK225083.1 - 1 209
BC003057.2 - 1 167
BC011713.2 - 1 197 3929724
BP342608.1 - 1 222

View Full Product Details