Gene Transcripts

Gene Symbol
WDR44
Entrez Gene ID
54521
Gene Name
WD repeat domain 44
Gene Aliases
RAB11BP, RPH11
Chromosome Location
Chr. X: 118345997 - 118449961 on Build GRCh38
UniGene ID
Hs.98510
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184965.1 NP_001171894.1 4 1224
NM_001184966.1 NP_001171895.1 3 1149
NM_019045.4 NP_061918.3 4 1224
XM_011531353.2 XP_011529655.1 3 1225
XM_017029600.1 XP_016885089.1 3 1157
GenBank mRNA AK092077.1 - 1 228
AK127556.1 - 4 1218
AK299711.1 - 3 1085
AK304432.1 - 4 751
AL833605.1 - 4 1189
AL834210.1 - 1 491
BC028697.2 - 4 1160 4837674
BX537578.1 - 4 1199
BX538087.1 - 4 1166
BX648150.1 - 4 1163

View Full Product Details