Gene Transcripts
- Gene Symbol
- LAS1L
- Entrez Gene ID
- 81887
- Gene Name
- LAS1 like, ribosome biogenesis factor
- Gene Aliases
- Las1-like, dJ475B7.2
- Chromosome Location
- Chr. X: 65512582 - 65534806 on Build GRCh38
- UniGene ID
- Hs.522675
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170649.1 |
NP_001164120.1
|
11, 12 |
1965 |
|
NM_001170650.1 |
NP_001164121.1
|
10, 11 |
1839 |
|
NM_031206.4 |
NP_112483.1
|
12, 13 |
2016 |
|
XM_011531045.1 |
XP_011529347.1
|
11, 12 |
1865 |
|
XM_017029879.1 |
XP_016885368.1
|
11, 12 |
1891 |
|
XM_017029880.1 |
XP_016885369.1
|
10, 11 |
1840 |
|
GenBank mRNA |
AF116730.1 |
- |
12, 13 |
1954 |
|
AK022587.1 |
- |
12, 13 |
1966 |
|
AK054817.1 |
- |
2, 3 |
1632 |
|
AK074087.1 |
- |
11, 12 |
4733 |
|
AK097383.1 |
- |
10, 11 |
1789 |
|
AK125760.1 |
- |
1, 2 |
1493 |
|
AK127207.1 |
- |
1, 2 |
1021 |
|
AK225059.1 |
- |
11, 12 |
1898 |
|
BC014545.1 |
- |
12, 13 |
2000 |
4079376 |
BC018610.1 |
- |
11, 12 |
1913 |
3140243 |
BC019302.1 |
- |
11, 12 |
1910 |
2822893 |
View Full Product Details