Gene Transcripts
- Gene Symbol
- TBC1D31
- Entrez Gene ID
- 93594
- Gene Name
- TBC1 domain family member 31
- Gene Aliases
- Gm85, WDR67
- Chromosome Location
- Chr. 8: 123041961 - 123165169 on Build GRCh38
- UniGene ID
- Hs.492716
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001145088.1 |
NP_001138560.1
|
1, 2 |
173 |
|
NM_145647.3 |
NP_663622.2
|
1, 2 |
173 |
|
XM_005251102.1 |
XP_005251159.1
|
1, 2 |
173 |
|
XM_005251103.1 |
XP_005251160.1
|
1, 2 |
173 |
|
XM_005251104.1 |
XP_005251161.1
|
1, 2 |
173 |
|
XM_011517379.2 |
XP_011515681.1
|
1, 2 |
173 |
|
XM_017013983.1 |
XP_016869472.1
|
1, 2 |
173 |
|
XM_017013985.1 |
XP_016869474.1
|
1, 2 |
284 |
|
XM_017013986.1 |
XP_016869475.1
|
1, 2 |
145 |
|
XM_017013987.1 |
XP_016869476.1
|
1, 2 |
173 |
|
XM_017013988.1 |
XP_016869477.1
|
1, 2 |
124 |
|
XM_017013989.1 |
XP_016869478.1
|
1, 2 |
287 |
|
XM_017013990.1 |
XP_016869479.1
|
1, 2 |
87 |
|
GenBank mRNA |
AK056434.1 |
- |
1, 2 |
174 |
|
AK094612.1 |
- |
1, 2 |
87 |
|
BC101724.1 |
- |
1, 2 |
140 |
8069230 |
BC105800.1 |
- |
1, 2 |
134 |
|
BC111954.1 |
- |
1, 2 |
140 |
8327422 |
BC143523.1 |
- |
1, 2 |
140 |
|
BC143525.1 |
- |
1, 2 |
140 |
9052034 |
View Full Product Details