Gene Transcripts
- Gene Symbol
- ANKRD18A
- Entrez Gene ID
- 253650
- Gene Name
- ankyrin repeat domain 18A
- Gene Aliases
- Chromosome Location
- Chr. 9: 38566260 - 38620587 on Build GRCh38
- UniGene ID
- Hs.644630
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093059.1 |
- |
8 |
2096 |
|
- Gene Symbol
- FAM95C
- Entrez Gene ID
- 100289137
- Gene Name
- family with sequence similarity 95 member C
- Gene Aliases
- Chromosome Location
- Chr. 9: 38540567 - 38545372 on Build GRCh38
- UniGene ID
- Hs.644630
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL834388.1 |
- |
1 |
593 |
|
View Full Product Details