Gene Transcripts
- Gene Symbol
- RSPH10B
- Entrez Gene ID
- 222967
- Gene Name
- radial spoke head 10 homolog B
- Gene Aliases
- Chromosome Location
- Chr. 7: 5926136 - 5970850 on Build GRCh38
- UniGene ID
- Hs.442339
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173565.3 |
NP_775836.3
|
12, 13 |
1682 |
|
XM_005249659.3 |
XP_005249716.1
|
12, 13 |
2184 |
|
XM_011515203.1 |
XP_011513505.1
|
13, 14 |
1779 |
|
XM_011515204.2 |
XP_011513506.1
|
12, 13 |
1967 |
|
XM_011515207.1 |
XP_011513509.1
|
12, 13 |
1659 |
|
XM_011515210.1 |
XP_011513512.1
|
6, 7 |
941 |
|
XM_011515211.1 |
XP_011513513.1
|
6, 7 |
814 |
|
XM_011515212.2 |
XP_011513514.1
|
12, 13 |
2199 |
|
XM_017011870.1 |
XP_016867359.1
|
12, 13 |
2199 |
|
GenBank mRNA |
BC034495.1 |
- |
12, 13 |
1685 |
|
BC157864.1 |
- |
10, 11 |
1432 |
9057083 |
BC157871.1 |
- |
10, 11 |
1432 |
9057090 |
- Gene Symbol
- RSPH10B2
- Entrez Gene ID
- 728194
- Gene Name
- radial spoke head 10 homolog B2
- Gene Aliases
- Chromosome Location
- Chr. 7: 6754109 - 6798775 on Build GRCh38
- UniGene ID
- Hs.710326
- Species
- Homo sapiens
View Full Product Details