Gene Transcripts
- Gene Symbol
- LY86-AS1
- Entrez Gene ID
- 285780
- Gene Name
- LY86 antisense RNA 1
- Gene Aliases
- LY86-AS, LY86AS
- Chromosome Location
- Chr. 6: 6346465 - 6622826 on Build GRCh38
- UniGene ID
- Hs.655737
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091027.1 |
- |
7 |
1160 |
|
View Full Product Details