Gene Transcripts
- Gene Symbol
- NKAIN3
- Entrez Gene ID
- 286183
- Gene Name
- Na+/K+ transporting ATPase interacting 3
- Gene Aliases
- FAM77D
- Chromosome Location
- Chr. 8: 62248942 - 62999652 on Build GRCh38
- UniGene ID
- Hs.654662
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_130764.1 |
|
6 |
1225 |
|
GenBank mRNA |
AK096949.1 |
- |
6 |
1225 |
|
View Full Product Details