Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173829.3 | NP_776190.1 | 5 | 586 | |
XM_011543344.2 | XP_011541646.1 | 4 | 718 | ||
GenBank mRNA | AK094073.1 | - | 586 | ||
AL832348.1 | - | 5 | 543 | ||
BC108716.1 | - | 5 | 517 | 6184559 | |
BC127085.1 | - | 5 | 524 | 40122236 | |
BC127086.1 | - | 5 | 524 | 40122239 |