Gene Transcripts

Gene Symbol
DCAF12L1
Entrez Gene ID
139170
Gene Name
DDB1 and CUL4 associated factor 12 like 1
Gene Aliases
KIAA1892L, WDR40B
Chromosome Location
Chr. X: 126549383 - 126552859 on Build GRCh38
UniGene ID
Hs.120403
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178470.4 NP_848565.2 1 1329
GenBank mRNA BC035674.1 - 1 1321

View Full Product Details