Gene Transcripts

Gene Symbol
THBS4
Entrez Gene ID
7060
Gene Name
thrombospondin 4
Gene Aliases
TSP-4, TSP4
Chromosome Location
Chr. 5: 79991296 - 80083287 on Build GRCh38
UniGene ID
Hs.211426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306212.1 NP_001293141.1 4 528
NM_001306213.1 NP_001293142.1 4 580
NM_001306214.1 NP_001293143.1 3 522
NM_003248.5 NP_003239.2 3 694
XM_017009798.1 XP_016865287.1 3 674
XM_017009799.1 XP_016865288.1 3 674
XM_017009800.1 XP_016865289.1 3 674
GenBank mRNA AK302842.1 - 3 522
AK313587.1 - 3 510
BC050456.1 - 4 778 6191808
DC310687.1 - 4 508
DC342285.1 - 4 529
Z19585.1 - 3 482

View Full Product Details