Gene Transcripts

Gene Symbol
AQP12A
Entrez Gene ID
375318
Gene Name
aquaporin 12A
Gene Aliases
AQP-12, AQP12, AQPX2
Chromosome Location
Chr. 2: 240691845 - 240698483 on Build GRCh38
UniGene ID
Hs.534650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198998.2 NP_945349.1 4 1024
GenBank mRNA AB040748.1 - 4 1024

Gene Symbol
AQP12B
Entrez Gene ID
653437
Gene Name
aquaporin 12B
Gene Aliases
INSSA3
Chromosome Location
Chr. 2: 240676418 - 240685743 on Build GRCh38
UniGene ID
Hs.437167
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001102467.1 NP_001095937.1 3 1054
XM_011511671.2 XP_011509973.1 9 1767
XM_011511672.2 XP_011509974.1 8 1707
XM_011511673.2 XP_011509975.1 9 1698
XM_011511675.2 XP_011509977.1 7 1629
XM_011511676.2 XP_011509978.1 6 1439
XM_017004736.1 XP_016860225.1 8 1710
XM_017004737.1 XP_016860226.1 5 1272
GenBank mRNA BC041460.1 - 5 2953
BC139748.1 - 3 1054 40115528
BC157895.1 - 3 1047 8862714
BC171912.1 - 3 1047 9054566

View Full Product Details