Gene Transcripts

Gene Symbol
RAB4B-EGLN2
Entrez Gene ID
100529264
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
Gene Aliases
RERT-lncRNA
Chromosome Location
Chr. 19: 40778219 - 40808441 on Build GRCh38
UniGene ID
Hs.631539
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037791.1 11, 12 2220
GenBank mRNA AK291385.1 - 11, 12 2168

Gene Symbol
EGLN2
Entrez Gene ID
112398
Gene Name
egl-9 family hypoxia inducible factor 2
Gene Aliases
EIT6, HIF-PH1, HIFPH1, HPH-1, HPH-3, PHD1
Chromosome Location
Chr. 19: 40799143 - 40808441 on Build GRCh38
UniGene ID
Hs.515417
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053046.3 NP_444274.1 5, 6 1616
NM_080732.3 NP_542770.2 5, 6 1526
GenBank mRNA AJ310544.1 - 4, 5 1468
AK025396.1 - 1, 2 47
AK026863.1 - 2, 3 771
AK074408.1 - 5, 6 1455
AK098182.1 - 4, 5 2200
AL133009.1 - 2, 3 3274
AL832506.1 - 5, 6 1482
AY040565.1 - 4, 5 1460
BC001723.1 - 5, 6 1429
BC036051.1 - 5, 6 1530 5270945

View Full Product Details