Gene Transcripts

Gene Symbol
TMC2
Entrez Gene ID
117532
Gene Name
transmembrane channel like 2
Gene Aliases
C20orf145
Chromosome Location
Chr. 20: 2536607 - 2642185 on Build GRCh38
UniGene ID
Hs.352626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080751.2 NP_542789.2 20 3067
XM_005260660.3 XP_005260717.1 18 3261
GenBank mRNA AF417580.2 - 20 3067

View Full Product Details