Gene Transcripts

Gene Symbol
GINM1
Entrez Gene ID
116254
Gene Name
glycoprotein integral membrane 1
Gene Aliases
C6orf72, dJ12G14.2
Chromosome Location
Chr. 6: 149566392 - 149590931 on Build GRCh38
UniGene ID
Hs.438872
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138785.3 NP_620140.1 8 956
GenBank mRNA AK289402.1 - 8 943
AK315729.1 - 8 953
AY358952.1 - 8 1074
BC014320.1 - 8 960 3686673

View Full Product Details