Gene Transcripts

Gene Symbol
TTC16
Entrez Gene ID
158248
Gene Name
tetratricopeptide repeat domain 16
Gene Aliases
Chromosome Location
Chr. 9: 127716064 - 127731600 on Build GRCh38
UniGene ID
Hs.642748
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317037.1 NP_001303966.1 5 559
NM_144965.2 NP_659402.1 5 598
XM_005251733.1 XP_005251790.1 4 410
XM_005251734.1 XP_005251791.1 5 583
XM_006716970.1 XP_006717033.1 5 583
XM_006716971.1 XP_006717034.1 4 410
XM_011518283.1 XP_011516585.1 5 596
XM_011518284.1 XP_011516586.1 4 454
XM_011518285.1 XP_011516587.1 5 596
XM_011518288.1 XP_011516590.1 4 454
XM_011518289.2 XP_011516591.1 5 507
XM_011518290.1 XP_011516592.1 4 454
XM_011518291.1 XP_011516593.1 5 583
XM_011518292.1 XP_011516594.1 4 454
XM_011518293.1 XP_011516595.1 5 596
XM_011518294.2 XP_011516596.1 5 596
XM_011518295.1 XP_011516597.1 5 596
XM_011518297.1 XP_011516599.1 5 596
XM_011518298.2 XP_011516600.1 5 596
GenBank mRNA AK057342.1 - 5 583
AK128011.1 - 2 525
AK294862.1 - 4 454
AK302422.1 - 4 410
AK302737.1 - 5 544
BC031281.1 - 5 596 5295890

View Full Product Details