Gene Transcripts

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
Chromosome Location
Chr. 10: 93667883 - 93702572 on Build GRCh38
UniGene ID
Hs.586650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145246.4 NP_660289.2 14 1860
XM_017015656.1 XP_016871145.1 15 1899
XM_017015657.1 XP_016871146.1 15 1934
XM_017015658.1 XP_016871147.1 16 2034
XM_017015659.1 XP_016871148.1 15 2126
XM_017015660.1 XP_016871149.1 16 1991
GenBank mRNA AK090955.1 - 14 1860

View Full Product Details