Gene Transcripts

Gene Symbol
PXT1
Entrez Gene ID
222659
Gene Name
peroxisomal, testis specific 1
Gene Aliases
STEPP
Chromosome Location
Chr. 6: 36390551 - 36442889 on Build GRCh38
UniGene ID
Hs.520097
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152990.3 NP_694535.2 5 1120
XM_011514400.2 XP_011512702.1 4 793
GenBank mRNA AF486827.1 - 2 417
BC031105.1 - 5 1121 5297409

View Full Product Details