Gene Transcripts

Gene Symbol
TSSK4
Entrez Gene ID
283629
Gene Name
testis specific serine kinase 4
Gene Aliases
C14orf20, STK22E, TSK-4, TSK4, TSSK5
Chromosome Location
Chr. 14: 24205524 - 24208362 on Build GRCh38
UniGene ID
Hs.314432
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184739.1 NP_001171668.1 4 1181
NM_001308067.1 NP_001294996.1 4 876
NM_174944.3 NP_777604.2 4 1151
XM_011536663.2 XP_011534965.1 4 1033
XM_017021226.1 XP_016876715.1 4 1419
GenBank mRNA AF542390.1 - 2 1144
AK131042.1 - 4 1181
AY461663.1 - 4 1115
BC111088.1 - 4 813 40036837
BI825382.1 - 2 223

View Full Product Details